siRNA Details
| Gene Symbol: Slc10a7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009981.2 | NP_001009981.1 | 2 | 397 | ||
| NM_001282108.1 | NP_001269037.1 | 2 | 397 | |||
| NM_001282109.1 | NP_001269038.1 | 2 | 397 | |||
| NM_029736.2 | NP_084012.1 | 2 | 397 | |||
| GenBank mRNA | AK010834.1 | - | 2 | 347 | ||
| AK040517.1 | - | 2 | 276 | |||
| AK079901.1 | - | 2 | 374 | |||
| AK152752.1 | - | 2 | 358 | |||
| AK152849.1 | - | 2 | 358 | |||
| AK165983.1 | - | 2 | 334 | |||
| AY825926.1 | - | 2 | 150 | |||
| AY825928.1 | - | 2 | 150 | |||
| BC116666.1 | - | 2 | 263 | 40107077 | ||
| BC116717.1 | - | 2 | 263 | 40107078 | ||

Slc10a7
Genomic Map