siRNA Details
| Gene Symbol: Cluap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029738.2 | NP_084014.1 | 9 | 1000 | ||
| GenBank mRNA | AK134648.1 | - | 9 | 967 | ||
| BC024910.1 | - | 9 | 1000 | 3603454 | ||
| BC063769.1 | - | 3 | 234 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cluap1| Gene Symbol: Cluap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029738.2 | NP_084014.1 | 9 | 1000 | ||
| GenBank mRNA | AK134648.1 | - | 9 | 967 | ||
| BC024910.1 | - | 9 | 1000 | 3603454 | ||
| BC063769.1 | - | 3 | 234 | |||