siRNA Details
| Gene Symbol: Klhdc10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029742.2 | NP_084018.1 | 5 | 762 | ||
| GenBank mRNA | AK010783.1 | - | 5 | 627 | ||
| AK039732.1 | - | 4 | 664 | |||
| AK050289.1 | - | 3 | 383 | |||
| AK053834.1 | - | 4 | 634 | |||
| AK085116.1 | - | 4 | 713 | |||
| AK085130.1 | - | 5 | 720 | |||
| AK172923.1 | - | 4 | 675 | |||
| BC057064.1 | - | 1 | 713 | |||
| BC060132.1 | - | 5 | 762 | 6409751 | ||

Klhdc10
Genomic Map