siRNA Details
| Gene Symbol: Klhdc10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029742.2 | NP_084018.1 | 4 | 572 | ||
| GenBank mRNA | AK010783.1 | - | 4 | 437 | ||
| AK039732.1 | - | 3 | 474 | |||
| AK050289.1 | - | 2 | 193 | |||
| AK053834.1 | - | 3 | 444 | |||
| AK085116.1 | - | 3 | 523 | |||
| AK085130.1 | - | 4 | 530 | |||
| AK172923.1 | - | 3 | 485 | |||
| BC060132.1 | - | 4 | 572 | 6409751 | ||

Klhdc10
Genomic Map