siRNA Details
| Gene Symbol: 2410141K09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183119.3 | NP_898942.1 | 5 | 464 | ||
| GenBank mRNA | AK010804.1 | - | 466 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2410141K09Rik| Gene Symbol: 2410141K09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183119.3 | NP_898942.1 | 5 | 464 | ||
| GenBank mRNA | AK010804.1 | - | 466 | |||