siRNA Details
| Gene Symbol: Bri3bp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029752.2 | NP_084028.1 | 2 | 368 | ||
| GenBank mRNA | AK010810.1 | - | 2 | 311 | ||
| AK043288.1 | - | 2 | 310 | |||
| AK044565.1 | - | 2 | 256 | |||
| AK143538.1 | - | 2 | 327 | |||
| BB618776.1 | - | 2 | 358 | |||
| BC039941.1 | - | 2 | 326 | 4011957 | ||
| BY152144.1 | - | 2 | 370 | |||

Bri3bp
Genomic Map