siRNA Details
| Gene Symbol: Sdccag8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029756.3 | NP_084032.1 | 8 | 1132 | ||
| GenBank mRNA | AK012392.1 | - | 8 | 920 | ||
| AK017691.1 | - | 8 | 915 | |||
| AK051683.1 | - | 8 | 935 | |||
| AK132316.1 | - | 8 | 928 | |||
| AK133622.1 | - | 8 | 876 | |||
| AK136921.1 | - | 8 | 955 | |||
| BC016244.1 | - | 8 | 912 | 4507575 | ||
| BC025138.1 | - | 7 | 788 | 4483636 | ||

Sdccag8
Genomic Map