siRNA Details
| Gene Symbol: Sdccag8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029756.3 | NP_084032.1 | 8 | 1094 | ||
| GenBank mRNA | AK012392.1 | - | 8 | 882 | ||
| AK017691.1 | - | 8 | 877 | |||
| AK051683.1 | - | 8 | 897 | |||
| AK132316.1 | - | 8 | 890 | |||
| AK133622.1 | - | 8 | 838 | |||
| AK136921.1 | - | 8 | 917 | |||
| BC016244.1 | - | 8 | 874 | 4507575 | ||
| BC025138.1 | - | 7 | 750 | 4483636 | ||

Sdccag8
Genomic Map