siRNA Details
| Gene Symbol: Sdccag8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029756.3 | NP_084032.1 | 7 | 1029 | ||
| GenBank mRNA | AK017691.1 | - | 7 | 812 | ||
| AK051683.1 | - | 7 | 832 | |||
| AK132316.1 | - | 7 | 825 | |||
| AK133622.1 | - | 7 | 773 | |||
| AK136921.1 | - | 7 | 852 | |||
| BC016244.1 | - | 7 | 809 | 4507575 | ||
| BC025138.1 | - | 6 | 685 | 4483636 | ||

Sdccag8
Genomic Map