siRNA Details
| Gene Symbol: Fam49a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146119.1 | NP_001139591.1 | 7 | 735 | ||
| NM_029758.4 | NP_084034.2 | 8 | 808 | |||
| GenBank mRNA | AK010821.1 | - | 6 | 671 | ||
| AK047142.1 | - | 7 | 698 | |||
| BC049664.1 | - | 8 | 770 | 6532813 | ||
| BC052465.1 | - | 7 | 716 | 6834768 | ||

Fam49a
Genomic Map