siRNA Details
| Gene Symbol: Dtl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB095735.1 | - | 14 | 1925 | ||
| AB095736.1 | - | 14 | 1925 | |||
| AK054412.1 | - | 14 | 1915 | |||
| AK133177.1 | - | 14 | 1924 | |||
| AK154594.1 | - | 15 | 1967 | |||
| AK161401.1 | - | 11 | 1526 | |||
| AK164249.1 | - | 14 | 1925 | |||
| AK166351.1 | - | 14 | 1933 | |||
| AK169601.1 | - | 14 | 1950 | |||
| BC060208.1 | - | 14 | 1884 | |||

Dtl
Genomic Map