siRNA Details
| Gene Symbol: Dcun1d5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029775.2 | NP_084051.1 | 7 | 866 | ||
| GenBank mRNA | AK013933.1 | - | 7 | 884 | ||
| AK014737.1 | - | 7 | 908 | |||
| AK134862.1 | - | 913 | ||||
| BC020048.1 | - | 7 | 902 | |||
| BC020089.1 | - | 7 | 866 | 4013755 | ||
| BC049641.1 | - | 1055 | ||||

Dcun1d5
Genomic Map