siRNA Details
| Gene Symbol: Fam81a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029784.2 | NP_084060.1 | 8 | 1166 | ||
| GenBank mRNA | AK018282.1 | - | 8 | 1123 | ||
| AK038532.1 | - | 8 | 1125 | |||
| AK044090.1 | - | 8 | 1166 | |||
| AK046408.1 | - | 8 | 1059 | |||
| AK079541.1 | - | 8 | 1126 | |||
| BC025646.1 | - | 8 | 1192 | 5324986 | ||

Fam81a
Genomic Map