siRNA Details
| Gene Symbol: Golga1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290649.1 | NP_001277578.1 | 9, 10 | 1114 | ||
| NM_029793.2 | NP_084069.1 | 10, 11 | 1189 | |||
| GenBank mRNA | AK028516.1 | - | 10, 11 | 1109 | ||
| AK044987.1 | - | 9, 10 | 1599 | |||
| BC024637.1 | - | 10, 11 | 1111 | |||
| BC043090.1 | - | 10, 11 | 1054 | 6418353 | ||
| BC053068.1 | - | 9, 10 | 1042 | 6405595 | ||
| BC080733.1 | - | 9, 10 | 1114 | 30110235 | ||

Golga1
Genomic Map