siRNA Details
Gene Symbol: Arfip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029802.4 | NP_084078.3 | 3 | 346 | ||
GenBank mRNA | AK009100.1 | - | 3 | 284 | ||
AK154772.1 | - | 3 | 312 | |||
BC013794.1 | - | 3 | 257 | 2648132 | ||
BC022942.1 | - | 3 | 270 | 3583149 | ||
BC028310.1 | - | 3 | 244 | |||
BC034520.1 | - | 3 | 327 | 3964827 | ||
BC079603.1 | - | 1 | 842 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001004222, XM_006229933, BC079174.
Panther Classification:
Gene Ontology Categories:
