siRNA Details
| Gene Symbol: Hnrnpm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109913.1 | NP_001103383.1 | 2 | 314 | ||
| NM_029804.3 | NP_084080.1 | 2 | 314 | |||
| GenBank mRNA | AK011521.1 | - | 2 | 260 | ||
| AK051976.1 | - | 2 | 260 | |||
| AK133911.1 | - | 2 | 249 | |||
| AK145343.1 | - | 2 | 252 | |||
| AK159848.1 | - | 2 | 246 | |||
| AK166670.1 | - | 2 | 249 | |||
| AK168347.1 | - | 2 | 249 | |||
| AK169101.1 | - | 2 | 249 | |||
| AK171892.1 | - | 2 | 249 | |||
| AK209547.1 | - | 1 | 122 | |||
| AK220299.1 | - | 2 | 274 | |||
| BC017539.1 | - | 2 | 244 | |||
| BC035525.1 | - | 2 | 249 | |||
| BC054785.1 | - | 1716 | ||||
| BC065172.1 | - | 2 | 244 | 6415170 | ||

Hnrnpm
Genomic Map