siRNA Details
| Gene Symbol: Psapl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175249.3 | NP_780458.2 | 1 | 1348 | ||
| GenBank mRNA | AK009408.1 | - | 1 | 1336 | ||
| AK028455.1 | - | 1 | 1313 | |||
| BC119540.2 | - | 1 | 1313 | |||
| BC148306.2 | - | 1 | 1313 | |||

Psapl1
Genomic Map