siRNA Details
| Gene Symbol: St5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001326.1 | NP_001001326.1 | 4 | 1518 | ||
| NM_029811.2 | NP_084087.2 | 3 | 267 | |||
| GenBank mRNA | AK158270.1 | - | 4 | 1523 | ||
| BC051034.1 | - | 4 | 1699 | 5345299 | ||
| KC789645.1 | - | 4 | 1118 | |||

St5
Genomic Map