siRNA Details
| Gene Symbol: Ddrgk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029832.2 | NP_084108.1 | 5, 6 | 722 | ||
| GenBank mRNA | BC051541.1 | - | 3, 4 | 311 | ||
| BC115616.2 | - | 5, 6 | 722 | 40102754 | ||
| BC115617.1 | - | 5, 6 | 722 | 40102757 | ||

Ddrgk1
Genomic Map