siRNA Details
| Gene Symbol: Tstd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029840.1 | NP_084116.1 | 2 | 200 | ||
| GenBank mRNA | AI287097.1 | - | 2 | 200 | ||
| AK011611.1 | - | 2 | 115 | |||
| AK154853.1 | - | 2 | 187 | |||
| AK218953.1 | - | 1 | 40 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tstd3| Gene Symbol: Tstd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029840.1 | NP_084116.1 | 2 | 200 | ||
| GenBank mRNA | AI287097.1 | - | 2 | 200 | ||
| AK011611.1 | - | 2 | 115 | |||
| AK154853.1 | - | 2 | 187 | |||
| AK218953.1 | - | 1 | 40 | |||