siRNA Details
| Gene Symbol: Kdm8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029842.5 | NP_084118.1 | 2 | 583 | ||
| GenBank mRNA | AK014006.1 | - | 2 | 527 | ||
| BC024807.1 | - | 2 | 511 | 5355270 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kdm8| Gene Symbol: Kdm8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029842.5 | NP_084118.1 | 2 | 583 | ||
| GenBank mRNA | AK014006.1 | - | 2 | 527 | ||
| BC024807.1 | - | 2 | 511 | 5355270 | ||