siRNA Details
Gene Symbol: 1700109H08Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029843.2 | NP_084119.1 | 2 | 214 | ||
GenBank mRNA | AK007150.1 | - | 2 | 257 | ||
BC048606.1 | - | 2 | 364 | |||
BC099581.1 | - | 2 | 214 | 30920581 |
