siRNA Details
| Gene Symbol: 1700109H08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029843.2 | NP_084119.1 | 2, 3 | 257 | ||
| GenBank mRNA | AK007150.1 | - | 2, 3 | 300 | ||
| BC048606.1 | - | 2, 3 | 407 | |||
| BC099581.1 | - | 2, 3 | 257 | 30920581 | ||

1700109H08Rik
Genomic Map