siRNA Details
| Gene Symbol: Arsk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029847.4 | NP_084123.2 | 4 | 603 | ||
| GenBank mRNA | AK013194.2 | - | 4 | 555 | ||
| AK019515.1 | - | 4 | 569 | |||
| AK031147.1 | - | 4 | 591 | |||
| AK032812.1 | - | 4 | 569 | |||
| AK033968.1 | - | 4 | 604 | |||
| AK035464.1 | - | 4 | 524 | |||
| AK039765.1 | - | 4 | 599 | |||
| AK046378.1 | - | 4 | 604 | |||
| AK083188.1 | - | 4 | 557 | |||
| AK084090.1 | - | 4 | 589 | |||
| AK086667.1 | - | 4 | 562 | |||
| AK139856.1 | - | 2 | 143 | |||
| AK144817.1 | - | 4 | 571 | |||
| AK165970.1 | - | 4 | 571 | |||
| BC046790.1 | - | 4 | 577 | 5717223 | ||
| BC058351.1 | - | 4 | 577 | 5717223 | ||

Arsk
Genomic Map