siRNA Details
Gene Symbol: Arid2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175251.4 | NP_780460.3 | 14 | 1957 | ||
GenBank mRNA | CA749551.1 | - | 6 | 706 | ||
CV559640.1 | - | 3 | 222 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Arid2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175251.4 | NP_780460.3 | 14 | 1957 | ||
GenBank mRNA | CA749551.1 | - | 6 | 706 | ||
CV559640.1 | - | 3 | 222 |