siRNA Details
| Gene Symbol: Cep83 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029852.2 | NP_084128.2 | 10, 11 | 1697 | ||
| GenBank mRNA | AK015007.1 | - | 10, 11 | 1834 | ||
| AK135868.1 | - | 2, 3 | 194 | |||
| AK151291.1 | - | 4, 5 | 546 | |||
| AK152465.1 | - | 4, 5 | 546 | |||
| AK152801.1 | - | 4, 5 | 546 | |||
| AK153515.1 | - | 4, 5 | 546 | |||
| BC028960.1 | - | 9, 10 | 1398 | |||
| BC043468.1 | - | 9, 10 | 1311 | |||
| BC070464.1 | - | 10, 11 | 1697 | 30608497 | ||

Cep83
Genomic Map