siRNA Details
Gene Symbol: 4921528I07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK014978.1 | - | 6, 7 | 399 | ||
AK077029.1 | - | 6, 7 | 399 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4921528I07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK014978.1 | - | 6, 7 | 399 | ||
AK077029.1 | - | 6, 7 | 399 |