siRNA Details
Gene Symbol: 4921528I07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK014978.1 | - | 5, 6 | 367 | ||
AK077029.1 | - | 5, 6 | 367 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4921528I07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK014978.1 | - | 5, 6 | 367 | ||
AK077029.1 | - | 5, 6 | 367 |