siRNA Details
Gene Symbol: Ccdc183 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029859.1 | NP_084135.1 | 2 | 202 | ||
GenBank mRNA | AK014983.1 | - | 2 | 184 | ||
BY084081.1 | - | 2 | 203 | |||
CF105570.1 | - | 2 | 188 |
