siRNA Details
| Gene Symbol: Ints7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK034856.1 | - | 9 | 1189 | ||
| AK040344.1 | - | 7 | 967 | |||
| AK052103.1 | - | 7 | 917 | |||
| AK085336.1 | - | 8 | 1063 | |||
| AK085362.1 | - | 9 | 1228 | |||
| AK086462.1 | - | 9 | 1221 | |||
| BC054120.1 | - | 9 | 1174 | 6506658 | ||

Ints7
Genomic Map