siRNA Details
| Gene Symbol: 9230110F15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029863.1 | NP_084139.1 | 2 | 226 | ||
| GenBank mRNA | AK020329.1 | - | 2 | 227 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

9230110F15Rik| Gene Symbol: 9230110F15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029863.1 | NP_084139.1 | 2 | 226 | ||
| GenBank mRNA | AK020329.1 | - | 2 | 227 | ||