siRNA Details
| Gene Symbol: Mtmr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023858.3 | NP_076347.3 | 13 | 1632 | ||
| GenBank mRNA | AK133476.1 | - | 13 | 1632 | ||
| AK133889.1 | - | 12 | 1400 | |||
| AK220369.1 | - | 6 | 7228 | |||
| AY055832.1 | - | 13 | 2403 | |||
| BC027013.1 | - | 4 | 1209 | |||
| BC063050.1 | - | 14 | 1721 | 5716297 | ||

Mtmr2
Genomic Map