siRNA Details
| Gene Symbol: Mtmr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023858.3 | NP_076347.3 | 4 | 480 | ||
| GenBank mRNA | AF073880.1 | - | 4 | 346 | ||
| AK020065.1 | - | 5 | 552 | |||
| AK133476.1 | - | 4 | 480 | |||
| AK133889.1 | - | 3 | 248 | |||
| AK176943.1 | - | 4 | 298 | |||
| AK220369.1 | - | 1 | 1599 | |||
| AY055832.1 | - | 4 | 1251 | |||
| BC063050.1 | - | 5 | 569 | 5716297 | ||

Mtmr2
Genomic Map