siRNA Details
| Gene Symbol: 9130221H12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020285.1 | - | 1 | 640 | ||
| BC022632.1 | - | 2 | 860 | 4207058 | ||
| Gene Symbol: Zfp526 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK173308.1 | - | 2119 | |||

9130221H12Rik
Genomic Map