siRNA Details
| Gene Symbol: Ptgr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252625.1 | NP_001239554.1 | 5 | 740 | ||
| NM_001252626.1 | NP_001239555.1 | 6 | 812 | |||
| NM_029880.3 | NP_084156.2 | 5 | 683 | |||
| GenBank mRNA | AK007522.1 | - | 6 | 696 | ||
| AK021033.1 | - | 5 | 575 | |||
| AK036168.1 | - | 5 | 605 | |||
| AK040396.1 | - | 6 | 744 | |||
| AK145232.1 | - | 5 | 631 | |||
| AK159187.1 | - | 5 | 579 | |||
| AK159932.1 | - | 5 | 580 | |||
| AK168895.1 | - | 5 | 574 | |||
| BC021466.1 | - | 5 | 629 | 5068419 | ||

Ptgr2
Genomic Map