siRNA Details
| Gene Symbol: Yif1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110201.1 | NP_001103671.1 | 3 | 408 | ||
| NM_029887.3 | NP_084163.2 | 3 | 417 | |||
| GenBank mRNA | AK020444.1 | - | 3 | 388 | ||
| AK135529.1 | - | 386 | ||||
| BC109331.2 | - | 3 | 387 | 40051521 | ||
| BU921944.1 | - | 3 | 339 | |||
| BU953116.1 | - | 3 | 405 | |||

Yif1b
Genomic Map