siRNA Details
| Gene Symbol: Yif1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110201.1 | NP_001103671.1 | 2 | 337 | ||
| NM_029887.3 | NP_084163.2 | 2 | 346 | |||
| GenBank mRNA | AK020444.1 | - | 2 | 317 | ||
| AK135529.1 | - | 315 | ||||
| AV482229.1 | - | 2 | 335 | |||
| BC109331.2 | - | 2 | 316 | 40051521 | ||
| BU921944.1 | - | 2 | 268 | |||
| BU953116.1 | - | 2 | 334 | |||

Yif1b
Genomic Map