siRNA Details
| Gene Symbol: Zfp142 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029888.2 | NP_084164.1 | 7 | 3915 | ||
| GenBank mRNA | AB093218.1 | - | 7 | 4202 | ||
| AF332091.1 | - | 6 | 3203 | |||
| AF332092.1 | - | 6 | 3203 | |||
| AK052201.1 | - | 1 | 856 | |||
| AK132550.1 | - | 7 | 3916 | |||
| BC060118.1 | - | 7 | 3851 | 6406631 | ||

Zfp142
Genomic Map