siRNA Details
Gene Symbol: Nkrf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029891.2 | NP_084167.2 | 3 | 1576 | ||
GenBank mRNA | AK042690.1 | - | 3 | 1578 | ||
AK079123.1 | - | 1 | 357 | |||
BC095972.1 | - | 1 | 335 | |||
BC138828.1 | - | 3 | 1544 | 8861850 | ||
BC138829.1 | - | 3 | 1544 | 8861851 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
