siRNA Details
| Gene Symbol: Rab35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198163.1 | NP_937806.1 | 6 | 669 | ||
| GenBank mRNA | AB232631.1 | - | 6 | 509 | ||
| AK156576.1 | - | 6 | 660 | |||
| AK185540.1 | - | 2 | 48 | |||
| AK191987.1 | - | 2 | 48 | |||
| AK195760.1 | - | 2 | 48 | |||
| AK197903.1 | - | 2 | 48 | |||
| AK200288.1 | - | 48 | ||||
| AK201126.1 | - | 2 | 48 | |||
| AK207070.1 | - | 2 | 48 | |||
| BC030941.1 | - | 4 | 360 | |||
| BC056466.1 | - | 6 | 669 | 5717246 | ||

Rab35
Genomic Map