siRNA Details
| Gene Symbol: Esrp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176838.2 | NP_789808.1 | 1 | 366 | ||
| GenBank mRNA | AK088755.1 | - | 1 | 235 | ||
| AK168488.1 | - | 1 | 214 | |||
| BC031444.1 | - | 1 | 366 | 4222237 | ||
| BC096581.1 | - | 1 | 206 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001107423, XM_006255496, BC166804.

Esrp2
Genomic Map