siRNA Details
| Gene Symbol: Myh10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175260.2 | NP_780469.1 | 34 | 5226 | ||
| GenBank mRNA | AK019969.1 | - | 14 | 2306 | ||
| AK029236.1 | - | 34 | 5033 | |||
| AK122578.1 | - | 32 | 4498 | |||
| AK147567.1 | - | 34 | 5205 | |||
| BC059863.1 | - | 9 | 1510 | |||
| BC089011.1 | - | 34 | 5002 | 30620877 | ||
| BC098465.1 | - | 15 | 2473 | |||

Myh10
Genomic Map