siRNA Details
| Gene Symbol: Myh10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175260.2 | NP_780469.1 | 2, 3 | 641 | ||
| GenBank mRNA | AK029236.1 | - | 2, 3 | 448 | ||
| AK045066.1 | - | 2, 3 | 443 | |||
| AK045074.1 | - | 2, 3 | 443 | |||
| AK047340.1 | - | 2, 3 | 476 | |||
| AK052435.1 | - | 2, 3 | 446 | |||
| AK133946.1 | - | 1, 2 | 407 | |||
| AK147567.1 | - | 2, 3 | 620 | |||
| AK163604.1 | - | 2, 3 | 457 | |||
| BC089011.1 | - | 2, 3 | 417 | 30620877 | ||

Myh10
Genomic Map