siRNA Details
| Gene Symbol: Myh10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175260.2 | NP_780469.1 | 8 | 1111 | ||
| GenBank mRNA | AK029236.1 | - | 8 | 918 | ||
| AK045066.1 | - | 8 | 913 | |||
| AK045074.1 | - | 8 | 913 | |||
| AK052435.1 | - | 8 | 916 | |||
| AK122578.1 | - | 6 | 383 | |||
| AK133946.1 | - | 7 | 877 | |||
| AK147567.1 | - | 8 | 1090 | |||
| AK163604.1 | - | 8 | 927 | |||
| BC089011.1 | - | 8 | 887 | 30620877 | ||

Myh10
Genomic Map