siRNA Details
| Gene Symbol: Usp45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290425.1 | NP_001277354.1 | 2 | 132 | ||
| NM_152825.2 | NP_690038.1 | 2 | 132 | |||
| GenBank mRNA | AK019756.1 | - | 2 | 174 | ||
| AK040660.1 | - | 2 | 133 | |||
| AK088073.1 | - | 2 | 115 | |||
| AK162825.1 | - | 2 | 133 | |||
| BB213271.2 | - | 2 | 144 | |||
| BC027768.1 | - | 2 | 115 | 5354502 | ||

Usp45
Genomic Map