siRNA Details
| Gene Symbol: Adgrf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133776.2 | NP_598537.2 | 12 | 2337 | ||
| GenBank mRNA | AK019869.1 | - | 12 | 2341 | ||
| BC019217.1 | - | 12 | 2366 | 5065702 | ||
UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Gene Symbol: Adgrf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133776.2 | NP_598537.2 | 12 | 2337 | ||
| GenBank mRNA | AK019869.1 | - | 12 | 2341 | ||
| BC019217.1 | - | 12 | 2366 | 5065702 | ||
Adgrf1
Genomic Map