siRNA Details
| Gene Symbol: Adgrf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133776.2 | NP_598537.2 | 12 | 2454 | ||
| GenBank mRNA | AK019869.1 | - | 12 | 2458 | ||
| AK052799.1 | - | 1 | 125 | |||
| BB661727.1 | - | 1 | 123 | |||
| BC019217.1 | - | 12 | 2483 | 5065702 | ||

Adgrf1
Genomic Map