siRNA Details
| Gene Symbol: Sphkap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172430.3 | NP_766018.3 | 5 | 545 | ||
| GenBank mRNA | AK020841.1 | - | 6 | 520 | ||
| AK034183.1 | - | 6 | 588 | |||
| AK038638.1 | - | 6 | 590 | |||
| BC069832.1 | - | 5 | 511 | 30431760 | ||

Sphkap
Genomic Map