siRNA Details
| Gene Symbol: Sphkap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172430.3 | NP_766018.3 | 7 | 1079 | ||
| GenBank mRNA | AK034183.1 | - | 8 | 1122 | ||
| AK038638.1 | - | 8 | 1124 | |||
| AK122538.1 | - | 1 | 29 | |||
| BC069832.1 | - | 7 | 1045 | 30431760 | ||

Sphkap
Genomic Map