siRNA Details
| Gene Symbol: NFKBIB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243116.1 | NP_001230045.1 | 6 | 846 | ||
| NM_002503.4 | NP_002494.2 | 6 | 1088 | |||
| NR_040515.1 | 5 | 2103 | ||||
| XM_006723226.2 | XP_006723289.2 | 6 | 1024 | |||
| XM_006723227.3 | XP_006723290.3 | 6 | 987 | |||
| GenBank mRNA | AA044256.1 | - | 1 | 223 | ||
| AK057862.1 | - | 5 | 2054 | |||
| BC015528.1 | - | 6 | 1019 | 3903794 | ||
| BT006743.1 | - | 6 | 985 | |||
| BX111397.1 | - | 1 | 229 | |||

NFKBIB
Genomic Map